accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5799-68-8

IUPAC Name: methanedisulfonyl dichloride
Synonyms: methanedisulfonyl dichloride
Molecular Weight (g/mol): 213.04
Molecular Formula: CH2Cl2O4S2
InChi Key: DKZQZHQHDMVGHE-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)CS(Cl)(=O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results