Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanedisulfonyl Dichloride 96.0+%, TCI America™
SDP

Catalog No. M16695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16695G 5 g
M166925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16695G Supplier TCI America Supplier No. M16695G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5799-68-8
Molecular Formula CH2Cl2O4S2
Molecular Weight (g/mol) 213.04
MDL Number MFCD00176854
InChI Key DKZQZHQHDMVGHE-UHFFFAOYSA-N
Synonym Methylenedisulfonyl Dichloride, Methanedisulfonyl Chloride
PubChem CID 79863
IUPAC Name methanedisulfonyl dichloride
SMILES ClS(=O)(=O)CS(Cl)(=O)=O

Specifications

Boiling Point 138°C
Quantity 5 g
UN Number 3265
Formula Weight 213.04
Percent Purity ≥96.0% (T)
Physical Form Liquid
Chemical Name or Material Methanedisulfonyl Dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.