accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58-71-9

IUPAC Name: sodium (6R,7R)-3-[(acetyloxy)methyl]-8-oxo-7-[2-(thiophen-2-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Synonyms: cephalothin sodium sodium cephalothin sodium cephalotin cefalotina sodica sodium cefalotin cefalotin sodium
Molecular Weight (g/mol): 418.41
Molecular Formula: C16H15N2NaO6S2
InChi Key: VUFGUVLLDPOSBC-XRZFDKQNSA-M
SMILES: [Na+].CC(=O)OCC1=C(N2[C@H](SC1)[C@H](NC(=O)CC1=CC=CS1)C2=O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)
Type 
  • (4)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)