accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOSNHOHN

CAS RN 58-85-5

IUPAC Name: 5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoic acid
Synonyms: -(+)-biotin d-biotin factor S
Molecular Weight (g/mol): 244.31
Molecular Formula: C10H16N2O3S
InChi Key: YBJHBAHKTGYVGT-UHFFFAOYNA-N
SMILES: OC(=O)CCCCC1SCC2NC(=O)NC12
Quantity 
  • (8)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (8)
  • (2)
  • (5)
Percent Purity 
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (6)
  • (7)
  • (4)
Physical Form 
  • (10)
  • (13)
  • (1)
  • (1)
Grade 
  • (3)
  • (1)
  • (6)
  • (1)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (22)
  • (13)
  • (1)
  • (2)
  • (7)
  • (13)
  • (3)
  • (8)
  • (12)
  • (8)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (8)
  • (2)
  • (5)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (6)
  • (7)
  • (4)
  • (10)
  • (13)
  • (1)
  • (1)
  • (3)
  • (1)
  • (6)
  • (1)
  • (7)
  • (2)
  • (1)