Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Biotin, MP Biomedicals™

Catalog No. MP021946345 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021946345 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021946345 Supplier MP Biomedicals, Inc Supplier No. 02194634.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Biotin is a growth factor present in small amounts in every living cell. It is involved in naturally occurring carboxylation reactions.

D-biotin occurs mainly bound to proteins or polypeptides. It is more abundant in the liver, kidney, pancreas, yeast and milk. Biotin levels are higher in cancerous tumors than in normal tissues. It is inactivated by binding to avidin.

  • Product is an essential vitamin that is important for amino acid and energy metabolism, and fatty acid synthesis.
  • It is a prosthetic group in four mammalian carboxylase families and facilitates the binding and transfer of carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-85-5
Molecular Formula C10H16N2O3S
Molecular Weight (g/mol) 244.31
MDL Number MFCD00005541
InChI Key YBJHBAHKTGYVGT-UHFFFAOYNA-N
Synonym biotin, d-biotin, vitamin h, coenzyme r, vitamin b7, bios ii, factor s, bioepiderm, d +-biotin, biodermatin
PubChem CID 171548
ChEBI CHEBI:15956
IUPAC Name 5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoic acid
SMILES OC(=O)CCCCC1SCC2NC(=O)NC12

Specifications

Melting Point 223°C to 233°C
Density 1.267 g/cm3(Lit.)
Boiling Point 573.6°C (760 mmHg)
Refractive Index 90.5° (C=2, 0.1mol/L NaOH)(Lit.)
Quantity 500 mg
Solubility Information Soluble in water (22 mg/100 mL), ethanol (80 mg/100 mL), more soluble in hot water and in dilute alkalies; insoluble in other common organic solvents. Soluble in 2 M Ammonium hydroxide (50mg/mL; clear, colorless solution), dimethylformamide (1.7 mg/mL).
Optical Rotation +89 to +93° (c=2, 0.1 N NaOH)
Formula Weight 244.3g/mol
Percent Purity ≥97.5%
Grade Cell Culture Reagent
Physical Form Crystalline Powder
Show More Show Less
Recommended Storage Store at +4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.