accessibility menu, dialog, popup

UserName

Viewing profile as user:
HON

CAS RN 580-16-5

IUPAC Name: quinolin-6-ol
Synonyms: 6-hydroxyquinoline 1H-1,6-epoxyquinoline 6-hdroxyquinoline 6-chinolinol
Molecular Weight (g/mol): 145.16
Molecular Formula: C9H7NO
InChi Key: OVYWMEWYEJLIER-UHFFFAOYSA-N
SMILES: OC1=CC=C2N=CC=CC2=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)