Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Hydroxyquinoline, MP Biomedicals

Catalog No. ICN215574 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN215574 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN215574 Supplier MP Biomedicals, Inc Supplier No. 0521557405

Chemical Identifiers

CAS 580-16-5
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
InChI Key OVYWMEWYEJLIER-UHFFFAOYSA-N
Synonym 6-hydroxyquinoline, 6-quinolinol, 1h-1,6-epoxyquinoline, ccris 4331, 6-chinolinol, 6-hydroxyquinoline 6-quinolinol, quinoline-6-ol, 6-quinolinoloxine, 6-hydroxy-chinolin, 6-hydroxy-quinoline
PubChem CID 11374
ChEBI CHEBI:48994
IUPAC Name quinolin-6-ol
SMILES C1=CC2=C(C=CC(=C2)O)N=C1

Specifications

Quantity 5 g
Formula Weight 145.16
Chemical Name or Material 6-Hydroxyquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.