accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 580-35-8

IUPAC Name: 2,4,6-triphenylpyridine
Synonyms: 2,4,6-triphenylpyridine
Molecular Weight (g/mol): 307.4
Molecular Formula: C23H17N
InChi Key: FRZHWQQBYDFNTH-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=CC(=NC(=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results