CAS RN 58002-62-3
IUPAC Name:
3-(carboxymethyl)-4-(prop-1-en-2-yl)pyrrolidine-2-carboxylic acid hydrate
Synonyms:
kainic acid hydrate
Molecular Weight (g/mol):
231.25
Molecular Formula:
C10H17NO5
InChi Key:
FZNZRJRSYLQHLT-UHFFFAOYNA-N
SMILES:
O.CC(=C)C1CNC(C1CC(O)=O)C(O)=O
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Kainic acid monohydrate, 96%
Thermo Scientific Chemicals Kainic acid monohydrate, 96%
CAS: 58002-62-3 Molecular Formula: C10H15NO4 Synonym: 2-Carboxy-3-carboxymethyl-4-isopropenylpyrrolidine; (-)-(α)-Kainic Acid