accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHOH2NOHH2OHClHCl

CAS RN 58052-48-5

IUPAC Name: 4-(aminomethyl)-5-(hydroxymethyl)-2-methylpyridin-3-ol hydrate dihydrochloride
Synonyms: pyridoxamine hydrate dihydrochloride
Molecular Weight (g/mol): 259.13
Molecular Formula: C8H16Cl2N2O3
InChi Key: UUEYKKGETIFWDR-UHFFFAOYSA-N
SMILES: O.Cl.Cl.CC1=NC=C(CO)C(CN)=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results