Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridoxamine Dihydrochloride Monohydrate 98.0+%, TCI America™
SDP

Catalog No. P05605G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P05605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P05605G Supplier TCI America Supplier No. P05605G

Specifications

CAS 58052-48-5
Melting Point 227°C
Color White-Yellow
Molecular Formula C8H16Cl2N2O3
Quantity 5 g
MDL Number MFCD00150098
InChI Key UUEYKKGETIFWDR-UHFFFAOYSA-N
SMILES CC1=NC=C(C(=C1O)CN)CO.O.Cl.Cl
IUPAC Name 4-(aminomethyl)-5-(hydroxymethyl)-2-methylpyridin-3-ol;hydrate;dihydrochloride
Molecular Weight (g/mol) 259.127
PubChem CID 21954278
Formula Weight 241.11
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Pyridoxamine Dihydrochloride Monohydrate
Show More Show Less
RTECSNumber UV1230000
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.