accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5809-91-6

IUPAC Name: ethenyl tetradecanoate
Synonyms: ethenyl tetradecanoate
Molecular Weight (g/mol): 254.41
Molecular Formula: C16H30O2
InChi Key: ZQZUENMXBZVXIZ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC(=O)OC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results