Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyl Myristate (stabilized with MEHQ) 99.0+%, TCI America™
SDP

Catalog No. M0723500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0723500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M0723500ML Supplier TCI America Supplier No. M0723500ML

missing translation for 'validMainframeMsg'

Specifications

CAS 5809-91-6
Color Colorless
Boiling Point 148°C
Molecular Formula C16H30O2
MDL Number MFCD00059221
Quantity 500 mL
Synonym Myristic Acid Vinyl Ester
InChI Key ZQZUENMXBZVXIZ-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCC(=O)OC=C
IUPAC Name ethenyl tetradecanoate
Molecular Weight (g/mol) 254.414
PubChem CID 545633
Formula Weight 254.41
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyl Myristate (stabilized with MEHQ)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.