accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOO

CAS RN 581-96-4

IUPAC Name: 2-(naphthalen-2-yl)acetic acid
Synonyms: β-naphthaleneacetic acid 2-naphthylacetic acid acide β-naphthylacetique betoxan
Molecular Weight (g/mol): 186.21
Molecular Formula: C12H10O2
InChi Key: VIBOGIYPPWLDTI-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C2C=CC=CC2=C1
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (2)
  • (7)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)