Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Naphthaleneacetic acid

Catalog No. N40025GA Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N40025GA 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N40025GA Supplier Sigma Aldrich Supplier No. N40025GA
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Physical properties (density, refractive index and boiling point) of trans,trans-farnesyl chloride have been reported.trans,trans-Farnesyl chloride (all-trans-farnesyl chloride) is suitable for use in the preparation of (S)-farnesyl thiopyrophosphate.

Specifications

CAS 581-96-4
Melting Point 141°C to 143°C (lit.)
Molecular Formula C12H10O2
Linear Formula C10H7CH2CO2H
MDL Number MFCD00004126
Quantity 5 g
Synonym 2-Naphthylacetic acid
Molecular Weight (g/mol) 186.21
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.