accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58117-53-6

IUPAC Name: benzyl N-[(5S)-5-amino-5-carbamoylpentyl]carbamate hydrochloride
Synonyms: benzyl N-[(5S)-5-amino-5-carbamoylpentyl]carbamate hydrochloride
Molecular Weight (g/mol): 315.8
Molecular Formula: C14H22ClN3O3
InChi Key: QSRGTPRDCDSKAY-YDALLXLXSA-N
SMILES: Cl.N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(N)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)