accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONH2O

CAS RN 5813-86-5

IUPAC Name: 3-methoxybenzamide
Synonyms: 3-methoxybenzamide M-methoxybenzamide
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: VKPLPDIMEREJJF-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results