accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58131-47-8

CH3SOOOCH3SOOOCa

CAS RN 58131-47-8

IUPAC Name: calcium dimethanesulfonate
Synonyms: calcium dimesylate
Molecular Weight (g/mol): 230.26
Molecular Formula: C2H6CaO6S2
InChi Key: BWEYVLQUNDGUEC-UHFFFAOYSA-L
SMILES: [Ca++].CS([O-])(=O)=O.CS([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results