accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5814-06-2

IUPAC Name: 2,4-dichlorobenzohydrazide
Synonyms: 2,4-dichlorobenzohydrazide
Molecular Weight (g/mol): 205.04
Molecular Formula: C7H6Cl2N2O
InChi Key: QOJQHOGSXXSMKX-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=C(Cl)C=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results