Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorobenzohydrazide 98.0+%, TCI America™
SDP

Catalog No. D48675G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48675G 5 g
D48671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48675G Supplier TCI America Supplier No. D48675G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5814-06-2
Molecular Formula C7H6Cl2N2O
Molecular Weight (g/mol) 205.04
MDL Number MFCD00025113
InChI Key QOJQHOGSXXSMKX-UHFFFAOYSA-N
Synonym 2,4-dichlorobenzhydrazide, 2,4-dichlorobenzoic acid hydrazide, 2,4-dichlorobenzenecarbohydrazide, acmc-1akyd, 2,4-dichlorobenzoylhydrazine, 2,4-dichlor-benzoyl-hydrazin, 2,4-dichlorobenzoic hydrazide, benzoic acid,2,4-dichloro-, hydrazide, benzoic acid, 2,4-dichloro-, hydrazide
PubChem CID 138596
IUPAC Name 2,4-dichlorobenzohydrazide
SMILES C1=CC(=C(C=C1Cl)Cl)C(=O)NN

Specifications

Melting Point 169°C
Color White
Quantity 5 g
Formula Weight 205.04
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichlorobenzohydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.