CAS RN 58161-35-6
CAS RN 58161-35-6
IUPAC Name:
N-(1-oxo-2,3-dihydro-1H-inden-5-yl)acetamide
Synonyms:
N-(1-oxo-2,3-dihydroinden-5-yl)acetamide
Molecular Weight (g/mol):
189.21
Molecular Formula:
C11H11NO2
InChi Key:
GHUPGGYDRVSZSW-UHFFFAOYSA-N
SMILES:
CC(=O)NC1=CC=C2C(=O)CCC2=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals N1-(1-Oxo-2,3-dihydro-1H-inden-5-yl)acetamide, 97%, Thermo Scientific™
CAS: 58161-35-6 Molecular Formula: C11H11NO2 Molecular Weight (g/mol): 189.214 MDL Number: MFCD00099465 InChI Key: GHUPGGYDRVSZSW-UHFFFAOYSA-N Synonym: n-1-oxo-2,3-dihydro-1h-inden-5-yl acetamide,n1-1-oxo-2,3-dihydro-1h-inden-5-yl acetamide,5-acetamido-1-indanone,5-acetylamino-1-indanone,n-1-oxoindan-5-yl acetamide,n-1-oxo-2,3-dihydroinden-5-yl acetamide,acetamide, n-2,3-dihydro-1-oxo-1h-inden-5-yl,5-acetamido-2,3-dihydro-1-oxo-1h-indene,5-acetamido-indanone PubChem CID: 312894 IUPAC Name: N-(1-oxo-2,3-dihydroinden-5-yl)acetamide SMILES: CC(=O)NC1=CC2=C(C=C1)C(=O)CC2