accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONHONHOO

CAS RN 58171-88-3

IUPAC Name: 2-(3-{[(benzyloxy)carbonyl]amino}propanamido)acetic acid
Synonyms: (3-{[(benzyloxy)carbonyl]amino}propanamido)acetic acid
Molecular Weight (g/mol): 280.28
Molecular Formula: C13H16N2O5
InChi Key: ILRSNXOKMYEWSE-UHFFFAOYSA-N
SMILES: OC(=O)CNC(=O)CCNC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results