Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyloxycarbonyl-beta-alanylglycine, 97%, Thermo Scientific™

Catalog No. AAH65424MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH65424MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH65424MD Supplier Thermo Scientific Chemicals Supplier No. H65424MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58171-88-3
Molecular Formula C13H16N2O5
Molecular Weight (g/mol) 280.28
MDL Number MFCD00055930
InChI Key ILRSNXOKMYEWSE-UHFFFAOYSA-N
Synonym 2-3-phenylmethoxycarbonylamino propanoylamino acetic acid, 3-benzyloxy carbonyl amino propanamido acetic acid, z-beta-ala-gly-oh, n-cbz-beta-alanylglycine, n-phenylmethoxy carbonyl-beta-alanylglycine, 2-3-benzyloxy carbonyl amino propanamido acetic acid, 3-benzyloxy carbonyl amino propanoyl amino acetic acid
PubChem CID 294919
IUPAC Name 2-[3-(phenylmethoxycarbonylamino)propanoylamino]acetic acid
SMILES C1=CC=C(C=C1)COC(=O)NCCC(=O)NCC(=O)O

Specifications

Beilstein 2008410
Quantity 250 mg
Formula Weight 280.28
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Benzyloxycarbonyl-beta-alanylglycine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.