accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OOO(?)

CAS RN 58239-72-8

IUPAC Name: 3-(16-methylheptadec-10-en-1-yl)oxolane-2,5-dione
Synonyms: 3-(16-methylheptadec-10-en-1-yl)oxolane-2,5-dione
Molecular Weight (g/mol): 350.54
Molecular Formula: C22H38O3
InChi Key: SJSHEXCFGWSYAD-VURMDHGXNA-N
SMILES: CC(C)CCCCC=CCCCCCCCCCC1CC(=O)OC1=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results