Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isooctadecenylsuccinic Anhydride (mixture of branched chain isomers) 90.0+%, TCI America™
SDP

Catalog No. I0237500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0237500G 500 g
I023725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0237500G Supplier TCI America Supplier No. I0237500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58239-72-8
Molecular Formula C22H38O3
Molecular Weight (g/mol) 350.54
MDL Number MFCD00070628
InChI Key SJSHEXCFGWSYAD-VURMDHGXNA-N

Specifications

Color Yellow
Quantity 500 g
Formula Weight 350.54
Percent Purity ≥90.0% (T)
Physical Form Liquid
Chemical Name or Material Isooctadecenylsuccinic Anhydride (mixture of branched chain isomers)
EINECSNumber (2)-0852
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.