accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHH2NHNNH2H2OH2OH2OH2OH2OH2OOHPOOHHOOHPOOHHO

CAS RN 58298-97-8

IUPAC Name: (3-aminopropyl)({4-[(3-aminopropyl)amino]butyl})amine bis(phosphoric acid) hexahydrate
Synonyms: bis(phosphoric acid) spermine hexahydrate
Molecular Weight (g/mol): 506.42
Molecular Formula: C10H44N4O14P2
InChi Key: WMFKFCJIERGVIW-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.OP(O)(O)=O.OP(O)(O)=O.NCCCNCCCCNCCCN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results