Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spermine Phosphate Hexahydrate 98.0+%, TCI America™
SDP

Catalog No. S00695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S00695G 5 g
S00691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S00695G Supplier TCI America Supplier No. S00695G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58298-97-8
Molecular Formula C10H26N4
MDL Number MFCD00050839
Synonym N,N′C-Bis(3-aminopropyl)-1,4-butanediamine Phosphate, N,N′C-Bis(3-aminopropyl)tetramethylenediamine Phosphate, Gerontine Phosphate, Musculamine Phosphate, Neuridine Phosphate

Specifications

Color White
Quantity 5 g
Formula Weight 398.33
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Spermine Phosphate Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.