accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 583-03-9

IUPAC Name: 1-phenylpentan-1-ol
Synonyms: α-butylbenzyl alcohol 1-phnp pancoral fenipentol, (S)- fenipentol, (R)- PH BC suiclisin (s)-isomer fenipentol fenipentol fenipentol, (s)-isomer
Molecular Weight (g/mol): 164.25
Molecular Formula: C11H16O
InChi Key: OVGORFFCBUIFIA-UHFFFAOYNA-N
SMILES: CCCCC(O)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)