Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-1-pentanol 98.0+%, TCI America™
SDP

Catalog No. P10865ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10865ML 5 mL
P108625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P10865ML Supplier TCI America Supplier No. P10865ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 583-03-9
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
MDL Number MFCD00021935
InChI Key OVGORFFCBUIFIA-UHFFFAOYSA-N
Synonym fenipentol, 1-phenyl-1-pentanol, 1-phenylpentanol, pancoral, phenylbutylcarbinol, suiclisin, alpha-butylbenzyl alcohol, 1-pentanol, 1-phenyl, 1-phenyl-1-hydroxypentane, ph bc
PubChem CID 3338
IUPAC Name 1-phenylpentan-1-ol
SMILES CCCCC(C1=CC=CC=C1)O

Specifications

Color Colorless
Boiling Point 137°C
Quantity 5 mL
Formula Weight 164.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1-pentanol
RTECSNumber DN8780000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.