accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O

CAS RN 583-60-8

IUPAC Name: 2-methylcyclohexan-1-one
Synonyms: 2-methylcyclohexanone
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: LFSAPCRASZRSKS-UHFFFAOYNA-N
SMILES: CC1CCCCC1=O
Molecular Weight (g/mol) 
  • (5)
  • (8)
Percent Purity 
  • (2)
  • (3)
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (11)
Boiling Point 
  • (3)
  • (2)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (9)
  • (5)
  • (8)
  • (2)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (11)
  • (3)
  • (2)
  • (3)
  • (2)