Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Methylcyclohexanone

Catalog No. M3840025ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M3840025ML 25 mL
M38400100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M3840025ML Supplier Sigma Aldrich Supplier No. M3840025ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Methylcyclohexanone

Specifications

CAS 583-60-8
Density 0.924 g/mL (at 25°C (literature))
Boiling Point 162°C to 163°C (lit.)
Molecular Formula C7H12O
Refractive Index n20/D 1.448 (literature)
Linear Formula CH3C6H9(=O)
MDL Number MFCD00001635
Quantity 25 mL
Synonym Tetrahydro-o-cresol
Molecular Weight (g/mol) 112.17
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.