accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 583-75-5

IUPAC Name: 4-bromo-2-methylaniline
Synonyms: benzenamine, 4-bromo-2-methyl-
Molecular Weight (g/mol): 186.05
Molecular Formula: C7H8BrN
InChi Key: PCHYYOCUCGCSBU-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CC=C1N
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)