Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-methylaniline 98.0+%, TCI America™
SDP

Catalog No. B128125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B128125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B128125G Supplier TCI America Supplier No. B128125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 583-75-5
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD00007825
InChI Key PCHYYOCUCGCSBU-UHFFFAOYSA-N
Synonym 2-amino-5-bromotoluene, 4-bromo-o-toluidine, 2-methyl-4-bromoaniline, benzenamine, 4-bromo-2-methyl, o-toluidine, 4-bromo, 4-bromo-2-methyl-phenylamine, 4-bromo-2-methylbenzenamine, 5-bromo-2-aminotoluene, 1-amino-4-bromo-2-methylbenzene, 4-bromo-2-methylbenenamine
PubChem CID 11423
IUPAC Name 4-bromo-2-methylaniline
SMILES CC1=CC(Br)=CC=C1N

Specifications

Melting Point 60°C
Boiling Point 240°C
Quantity 25 g
Formula Weight 186.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-methylaniline
RTECSNumber XU4250000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.