CAS RN 58333-75-8
IUPAC Name:
4-(2-methoxyphenyl)piperidine
Synonyms:
4-(2-methoxyphenyl)piperidine
Molecular Weight (g/mol):
191.27
Molecular Formula:
C12H17NO
InChi Key:
SRAVSVBVHDLLPO-UHFFFAOYSA-N
SMILES:
COC1=CC=CC=C1C1CCNCC1
1
–
1
of
1
results
1
4-(2-Methoxyphenyl)piperidine, 97%, Thermo Scientific™
CAS: 58333-75-8 Molecular Formula: C12H17NO Molecular Weight (g/mol): 191.274 MDL Number: MFCD00277219 InChI Key: SRAVSVBVHDLLPO-UHFFFAOYSA-N PubChem CID: 544738 IUPAC Name: 4-(2-methoxyphenyl)piperidine SMILES: COC1=CC=CC=C1C2CCNCC2