CAS RN 5837-73-0
IUPAC Name:
methyl 2-hydroxybut-3-enoate
Synonyms:
methyl 2-hydroxybut-3-enoate
Molecular Weight (g/mol):
116.12
Molecular Formula:
C5H8O3
InChi Key:
JBKTVPSFVUFSAO-UHFFFAOYNA-N
SMILES:
COC(=O)C(O)C=C
1
–
1
of
1
results
1
Methyl DL-2-Hydroxy-3-butenoate 98.0+%, TCI America™
CAS: 5837-73-0 Molecular Formula: C5H8O3 Molecular Weight (g/mol): 116.116 MDL Number: MFCD00144164 InChI Key: JBKTVPSFVUFSAO-UHFFFAOYSA-N Synonym: DL-2-Hydroxy-3-butenoic Acid Methyl Ester PubChem CID: 545123 IUPAC Name: methyl 2-hydroxybut-3-enoate SMILES: COC(=O)C(C=C)O