Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl DL-2-Hydroxy-3-butenoate 98.0+%, TCI America™
SDP

Catalog No. H07485G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H07485G Supplier TCI America Supplier No. H07485G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5837-73-0
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00144164
InChI Key JBKTVPSFVUFSAO-UHFFFAOYSA-N
Synonym DL-2-Hydroxy-3-butenoic Acid Methyl Ester
PubChem CID 545123
IUPAC Name methyl 2-hydroxybut-3-enoate
SMILES COC(=O)C(C=C)O

Specifications

Color Yellow
Boiling Point 64°C
Quantity 5 g
Formula Weight 116.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl DL-2-Hydroxy-3-butenoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.