accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 584-02-1

IUPAC Name: pentan-3-ol
Synonyms: 3-pentanol
Molecular Weight (g/mol): 88.15
Molecular Formula: C5H12O
InChi Key: AQIXEPGDORPWBJ-UHFFFAOYSA-N
SMILES: CCC(O)CC
Boiling Point 
  • (2)
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (6)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (5)