Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Pentanol

Catalog No. P8025500MLA Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P8025500MLA 500 mL
P802525MLA 25 mL
P8025100MLA 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P8025500MLA Supplier Sigma Aldrich Supplier No. P8025500MLA
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-pentanol can be used as:
  • A starting material for the preparation of liquid crystals, 1-ethylpropyl (r)-2-[4-(4α-alkoxybiphenylcarbonyloxy)-phenoxy]propionates by reacting with chiral (s)-lactic acid
  • Solvent/reductant in the catalytic deoxydehydration reaction of c4c6 sugar alcohols into linear polyene using methyltrioxorhenium as a catalyst
  • A reactant for the synthesis of 3-(4-bromophenyloxy)pentane by reacting with 4-bromophenol via base-catalyzed mitsunobu reaction

Specifications

CAS 584-02-1
Density 0.815 g/mL (at 25°C (literature))
Boiling Point 114°C to 115°C (749 mmHg)
Molecular Formula C5H12O
Refractive Index n20/D 1.410 (literature)
Linear Formula (C2H5)2 CHOH
MDL Number MFCD00004574
Quantity 500 mL
Synonym sec-inact.-Pentyl alcohol; Diethyl carbinol
Molecular Weight (g/mol) 88.15
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.