accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CBrOHO

CAS RN 584-93-0

IUPAC Name: 2-bromopentanoic acid
Synonyms: 2-bromopentanoic acid
Molecular Weight (g/mol): 181.03
Molecular Formula: C5H9BrO2
InChi Key: WMFATTFQNRPXBQ-UHFFFAOYNA-N
SMILES: CCCC(Br)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)