Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Bromo-n-valeric Acid, MP Biomedicals

Catalog No. ICN204801 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN204801 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN204801 Supplier MP Biomedicals, Inc Supplier No. 0520480110

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 584-93-0
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00004217
InChI Key WMFATTFQNRPXBQ-UHFFFAOYNA-N
Synonym 2-bromovaleric acid, pentanoic acid, 2-bromo, .alpha.-bromovaleric acid, valeric acid, 2-bromo, alpha-bromo-n-valeric acid, .alpha.-bromopentanoic acid, valeric acid, .alpha.-bromo, alpha-bromovaleric acid, alpha-bromopentanoic acid, valeric acid, alpha-bromo
PubChem CID 11446
IUPAC Name 2-bromopentanoic acid
SMILES CCCC(Br)C(O)=O

Specifications

Quantity 10 g
Formula Weight 181.04
Chemical Name or Material α-Bromo-n-valeric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.