accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 5840-01-7

IUPAC Name: 1-azatricyclo[6.3.1.0⁴,¹²]dodeca-2,4,6,8(12)-tetraene
Synonyms: 1-azatricyclo[6.3.1.0⁴,¹²]dodeca-2,4,6,8(12)-tetraene
Molecular Weight (g/mol): 157.22
Molecular Formula: C11H11N
InChi Key: QCCKSFHMARIKSK-UHFFFAOYSA-N
SMILES: C1CN2C=CC3=CC=CC(C1)=C23
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results