Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lilolidine, 98%, Thermo Scientific™

Catalog No. AC452900050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC452900050 5 g
AC452900010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC452900050 Supplier Thermo Scientific Chemicals Supplier No. 452900050
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5840-01-7
Molecular Formula C11H11N
Molecular Weight (g/mol) 157.21
InChI Key QCCKSFHMARIKSK-UHFFFAOYSA-N

Specifications

Loss on Drying 1% max. (60°C) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Synonym lilolidine, 5,6-dihydro-4h-pyrrolo 3,2,1-ij quinoline, 1,7-trimethyleneindole, 2,3-dihydro-1h-pyrrolo 3,2,1-ij quinoline, 4h-pyrrolo 3,2,1-ij quinoline, 5,6-dihydro, lilolidene, 1-azatricyclo 6.3.1.0?, 1 2 dodeca-2,4,6,8 12-tetraene, g00001-watson-int, ksc917k8l
SMILES C1CC2=CC=CC3=C2N(C1)C=C3
PubChem CID 15645629
Formula Weight 157.21
Percent Purity 98%
Chemical Name or Material Lilolidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.