accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrBrBrBr

CAS RN 58443-86-0

IUPAC Name: 1,2,5,6-tetrabromohexane
Synonyms: 1,2,5,6-tetrabromohexane
Molecular Weight (g/mol): 401.76
Molecular Formula: C6H10Br4
InChi Key: WPBWUVCMCYXPFI-UHFFFAOYNA-N
SMILES: BrCC(Br)CCC(Br)CBr
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)