Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,5,6-Tetrabromohexane (mixture of diastereoisomers) 97.0+%, TCI America™
SDP

Catalog No. T29835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29835G 5 g
T298325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29835G Supplier TCI America Supplier No. T29835G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 58443-86-0
Molecular Formula C6H10Br4
Molecular Weight (g/mol) 401.762
MDL Number MFCD00049137
InChI Key WPBWUVCMCYXPFI-UHFFFAOYSA-N
Synonym hexane, 1,2,5,6-tetrabromo, acmc-20ap74, hexane,1,2,5,6-tetrabromo
PubChem CID 94047
IUPAC Name 1,2,5,6-tetrabromohexane
SMILES C(CC(CBr)Br)C(CBr)Br

Specifications

Melting Point 49°C
Color White
Quantity 5 g
Formula Weight 401.76
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,5,6-Tetrabromohexane (mixture of diastereoisomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.