accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNNNNOHOOOOOO

CAS RN 58463-04-0

IUPAC Name: 5-[6-(N-benzoylbenzamido)-9H-purin-9-yl]-4-(benzoyloxy)-2-(hydroxymethyl)oxolan-3-yl benzoate
Synonyms: 5-[6-(N-benzoylbenzamido)purin-9-yl]-4-(benzoyloxy)-2-(hydroxymethyl)oxolan-3-yl benzoate
Molecular Weight (g/mol): 683.68
Molecular Formula: C38H29N5O8
InChi Key: GNYQOUZNLMTKQG-UHFFFAOYNA-N
SMILES: OCC1OC(C(OC(=O)C2=CC=CC=C2)C1OC(=O)C1=CC=CC=C1)N1C=NC2=C(N=CN=C12)N(C(=O)C1=CC=CC=C1)C(=O)C1=CC=CC=C1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)