accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5847-59-6

IUPAC Name: 2-bromo-4-nitrophenol
Synonyms: 2-bromo-4-nitrophenol
Molecular Weight (g/mol): 218.01
Molecular Formula: C6H4BrNO3
InChi Key: DCIPFSYBGTWYCR-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1Br)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)