Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-nitrophenol 98.0+%, TCI America™
SDP

Catalog No. B39185G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39185G 5 g
B39181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39185G Supplier TCI America Supplier No. B39185G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5847-59-6
Molecular Formula C6H4BrNO3
Molecular Weight (g/mol) 218.006
MDL Number MFCD06656567
InChI Key DCIPFSYBGTWYCR-UHFFFAOYSA-N
Synonym 3-Bromo-4-hydroxynitrobenzene
PubChem CID 22109
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Br)O

Specifications

Melting Point 112°C
Color Yellow
Quantity 5 g
UN Number 3077
Formula Weight 218.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-nitrophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.