accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58473-78-2

IUPAC Name: 4-(1-{4-[bis(4-methylphenyl)amino]phenyl}cyclohexyl)-N,N-bis(4-methylphenyl)aniline
Synonyms: 4-(1-{4-[bis(4-methylphenyl)amino]phenyl}cyclohexyl)-N,N-bis(4-methylphenyl)aniline
Molecular Weight (g/mol): 626.89
Molecular Formula: C46H46N2
InChi Key: ZOKIJILZFXPFTO-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)N(C1=CC=C(C)C=C1)C1=CC=C(C=C1)C1(CCCCC1)C1=CC=C(C=C1)N(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results