Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Bis[4-[N,N-di(p-tolyl)amino]phenyl]cyclohexane 98.0+%, TCI America™
SDP

Catalog No. B20795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20795G 5 g
B20791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20795G Supplier TCI America Supplier No. B20795G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 58473-78-2
Molecular Formula C46H46N2
Molecular Weight (g/mol) 626.888
MDL Number MFCD03844768
InChI Key ZOKIJILZFXPFTO-UHFFFAOYSA-N
PubChem CID 94071
IUPAC Name 4-methyl-N-[4-[1-[4-(4-methyl-N-(4-methylphenyl)anilino)phenyl]cyclohexyl]phenyl]-N-(4-methylphenyl)aniline
SMILES CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)C4(CCCCC4)C5=CC=C(C=C5)N(C6=CC=C(C=C6)C)C7=CC=C(C=C7)C

Specifications

Melting Point 186°C
Quantity 5 g
Synonym 4,4′-Cyclohexylidenebis[N,N-di(p-tolyl)aniline], TAPC
Formula Weight 626.89
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Bis[4-[N,N-di(p-tolyl)amino]phenyl]cyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.