CAS RN 58479-61-1
tert-Butylchlorodiphenylsilane, 98%
CAS: 58479-61-1 Molecular Formula: C16H19ClSi Molecular Weight (g/mol): 274.86 MDL Number: MFCD00000497 InChI Key: MHYGQXWCZAYSLJ-UHFFFAOYSA-N Synonym: tert-butylchlorodiphenylsilane,tert-butyldiphenylchlorosilane,t-butyldiphenylchlorosilane,silane, chloro 1,1-dimethylethyl diphenyl,tert-butyl chloro diphenylsilane,t-butylchlorodiphenylsilane,tbdpscl,t-butyldiphenylsilyl chloride,tert-butyldiphenyl chlorosilane,unii-3beu48ui4e PubChem CID: 94078 IUPAC Name: tert-butyl-chloro-diphenylsilane SMILES: CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl
tert-Butyldiphenylchlorosilane, 97%
CAS: 58479-61-1 Molecular Formula: C16H19ClSi Molecular Weight (g/mol): 274.863 MDL Number: MFCD00000497 InChI Key: MHYGQXWCZAYSLJ-UHFFFAOYSA-N Synonym: tert-butylchlorodiphenylsilane,tert-butyldiphenylchlorosilane,t-butyldiphenylchlorosilane,silane, chloro 1,1-dimethylethyl diphenyl,tert-butyl chloro diphenylsilane,t-butylchlorodiphenylsilane,tbdpscl,t-butyldiphenylsilyl chloride,tert-butyldiphenyl chlorosilane,unii-3beu48ui4e PubChem CID: 94078 IUPAC Name: tert-butyl-chloro-diphenylsilane SMILES: CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl
tert-Butylchlorodiphenylsilane, 98%, AcroSeal™
CAS: 58479-61-1 Molecular Formula: C16H19ClSi Molecular Weight (g/mol): 274.86 MDL Number: MFCD00000497 InChI Key: MHYGQXWCZAYSLJ-UHFFFAOYSA-N Synonym: tert-butylchlorodiphenylsilane,tert-butyldiphenylchlorosilane,t-butyldiphenylchlorosilane,silane, chloro 1,1-dimethylethyl diphenyl,tert-butyl chloro diphenylsilane,t-butylchlorodiphenylsilane,tbdpscl,t-butyldiphenylsilyl chloride,tert-butyldiphenyl chlorosilane,unii-3beu48ui4e PubChem CID: 94078 IUPAC Name: tert-butyl-chloro-diphenylsilane SMILES: CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl
tert-Butyldiphenylchlorosilane 97.0+%, TCI America™
CAS: 58479-61-1 Molecular Formula: C16H19ClSi Molecular Weight (g/mol): 274.863 MDL Number: MFCD00000497 InChI Key: MHYGQXWCZAYSLJ-UHFFFAOYSA-N Synonym: tert-butylchlorodiphenylsilane,tert-butyldiphenylchlorosilane,t-butyldiphenylchlorosilane,silane, chloro 1,1-dimethylethyl diphenyl,tert-butyl chloro diphenylsilane,t-butylchlorodiphenylsilane,tbdpscl,t-butyldiphenylsilyl chloride,tert-butyldiphenyl chlorosilane,unii-3beu48ui4e PubChem CID: 94078 IUPAC Name: tert-butyl-chloro-diphenylsilane SMILES: CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl
Sigma Aldrich 4-Tert-Butylphenylmagnesium Bromide Solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Tert-Butyl(Chloro)Diphenylsilane
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.